Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026794.2 | NP_081070.1 | 1, 2 | 83 | ||
GenBank mRNA | AK003863.1 | - | 1, 2 | 91 | ||
AK049702.1 | - | 1, 2 | 125 | |||
BC027548.1 | - | 1, 2 | 83 | 1532650 |