Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172980.2 | NP_766568.1 | 6 | 1726 | ||
GenBank mRNA | AF079853.2 | - | 5 | 601 | ||
AK080957.1 | - | 6 | 1727 | |||
AK152776.1 | - | 5 | 661 | |||
BC055376.1 | - | 5 | 620 | 30077988 |