Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012030.2 | NP_036160.1 | 1 | 473 | ||
GenBank mRNA | AK085210.1 | - | 1 | 474 | ||
AK153558.1 | - | 1 | 473 | |||
AK168894.1 | - | 1 | 474 | |||
BB660628.1 | - | 1 | 474 | |||
BC038656.1 | - | 1 | 345 | |||
BC085141.1 | - | 1 | 458 | 6413140 |