Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jade1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130184.1 | NP_001123656.1 | 11 | 2457 | ||
NM_001130185.1 | NP_001123657.1 | 11 | 2537 | |||
NM_001130186.1 | NP_001123658.1 | 11 | 2495 | |||
NM_172303.4 | NP_758507.3 | 11 | 2555 | |||
GenBank mRNA | AK049332.1 | - | 11 | 2460 | ||
AK129445.1 | - | 11 | 2450 | |||
AK147466.1 | - | 11 | 2417 | |||
BC020316.2 | - | 11 | 2477 | 5250231 | ||
BC026471.1 | - | 1 | 406 | |||
BC038026.1 | - | 3 | 1234 |