Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081323.2 | NP_001074792.1 | 9 | 1988 | ||
NM_001277867.1 | NP_001264796.1 | 10 | 2078 | |||
GenBank mRNA | AK081178.1 | - | 9 | 1837 | ||
AK084586.1 | - | 9 | 1822 | |||
BC076624.1 | - | 9 | 1851 | |||
BC138423.1 | - | 6 | 1390 | 8861443 | ||
BC145681.1 | - | 6 | 1390 | 40130984 |