Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252382.1 | NP_001239311.1 | 9 | 1691 | ||
NM_012057.4 | NP_036187.1 | 9 | 1639 | |||
GenBank mRNA | AF028725.1 | - | 9 | 1531 | ||
AK150598.1 | - | 9 | 1558 | |||
AK151019.1 | - | 9 | 1556 | |||
AK152042.1 | - | 9 | 1558 | |||
AK156220.1 | - | 9 | 1691 | |||
EU401974.1 | - | 6 | 1190 |