Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013785.2 | NP_038813.2 | 6 | 1338 | ||
GenBank mRNA | AF177144.1 | - | 5 | 1038 | ||
AK037925.1 | - | 5 | 1148 | |||
AK088872.1 | - | 6 | 1356 | |||
AK171310.1 | - | 6 | 1756 | |||
AK172921.1 | - | 6 | 1397 | |||
BC053030.1 | - | 6 | 1342 | |||
BC059006.1 | - | 6 | 1338 | 6827203 |