Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013791.2 | NP_038819.1 | 13 | 1604 | ||
GenBank mRNA | AK077541.1 | - | 13 | 1599 | ||
AK136123.1 | - | 10 | 1289 | |||
BC013703.1 | - | 13 | 1588 | 3668925 | ||
U72194.1 | - | 13 | 1587 |