Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700088E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138581.2 | NP_613047.1 | 2 | 167 | ||
GenBank mRNA | AA008475.1 | - | 2 | 156 | ||
AK006539.1 | - | 2 | 148 | |||
AK007028.1 | - | 2 | 174 | |||
AK030004.1 | - | 120 | ||||
BB602239.1 | - | 2 | 170 | |||
BC068203.1 | - | 1 | 212 | |||
BC096672.1 | - | 1 | 265 | 5117646 | ||
BC116891.1 | - | 2 | 98 | 40126210 | ||
BC116893.1 | - | 2 | 98 | 40126212 | ||
BC144714.1 | - | 2 | 98 | 9053262 | ||
BC144715.1 | - | 2 | 98 |