Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172151.4 | NP_742163.4 | 11 | 2389 | ||
GenBank mRNA | AK032069.1 | - | 2 | 379 | ||
AY668947.1 | - | 11 | 2263 | |||
AY894892.1 | - | 11 | 2389 | |||
BC004683.1 | - | 1 | 200 | |||
BC055694.1 | - | 2 | 686 | |||
BC083154.1 | - | 1 | 310 | |||
BC106166.1 | - | 1 | 292 |