Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178600.2 | NP_848715.1 | 2 | 303 | ||
GenBank mRNA | AK002742.1 | - | 2 | 284 | ||
AK003237.1 | - | 2 | 304 | |||
AK008509.1 | - | 2 | 277 | |||
AK008578.1 | - | 2 | 278 | |||
AK013996.1 | - | 2 | 287 | |||
BC031732.1 | - | 2 | 285 | 3991412 | ||
CD774913.1 | - | 2 | 458 |