Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128599.1 | NP_001122071.1 | 6 | 1075 | ||
NM_013887.2 | NP_038915.1 | 6 | 1075 | |||
GenBank mRNA | AF147789.1 | - | 6 | 1075 | ||
BC139827.1 | - | 6 | 1016 | 40142922 | ||
DQ278263.1 | - | 2 | 238 | |||
EU303117.1 | - | 6 | 891 | |||
EU303118.1 | - | 6 | 891 |
Set Membership: |
![]() |