Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128599.1 | NP_001122071.1 | 7 | 1218 | ||
NM_013887.2 | NP_038915.1 | 7 | 1218 | |||
GenBank mRNA | AF147789.1 | - | 7 | 1218 | ||
BC139827.1 | - | 7 | 1159 | 40142922 | ||
DQ278263.1 | - | 3 | 381 | |||
EU303117.1 | - | 7 | 1034 | |||
EU303118.1 | - | 7 | 1034 |
Set Membership: |
![]() |