Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013895.4 | NP_038923.1 | 3 | 484 | ||
GenBank mRNA | AF144702.1 | - | 3 | 486 | ||
AK003054.1 | - | 3 | 290 | |||
AK009041.1 | - | 3 | 278 | |||
AK009152.1 | - | 3 | 294 | |||
AK013230.1 | - | 3 | 282 | |||
AK088984.1 | - | 3 | 294 | |||
BC011436.1 | - | 3 | 297 | 2812494 |