Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024853.1 | NP_001020024.1 | 5 | 387 | ||
NM_001024854.1 | NP_001020025.1 | 3 | 203 | |||
NM_001286203.2 | NP_001273132.1 | 4 | 289 | |||
NM_013896.3 | NP_038924.1 | 5 | 425 | |||
GenBank mRNA | AF150101.1 | - | 4 | 248 | ||
AI119857.1 | - | 5 | 406 | |||
AK018764.1 | - | 3 | 203 | |||
AK165848.1 | - | 3 | 204 | |||
AK172179.1 | - | 4 | 299 | |||
BC024370.1 | - | 5 | 398 | 4976059 | ||
CK626183.1 | - | 3 | 185 | |||
CO799389.1 | - | 4 | 289 | |||
CO809703.1 | - | 356 | ||||
DV652207.1 | - | 5 | 364 |