Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h2be | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178214.3 | NP_835586.2 | 1 | 339 | ||
GenBank mRNA | AK078533.1 | - | 1 | 340 | ||
AK090189.1 | - | 1 | 342 | |||
AK143398.1 | - | 1 | 343 | |||
AK162494.1 | - | 1 | 343 | |||
AK167546.1 | - | 1 | 342 | |||
AK168767.1 | - | 1 | 339 | |||
AK170749.1 | - | 1 | 342 | |||
BC047137.1 | - | 1 | 319 | 6311571 | ||
BC058562.1 | - | 1 | 341 |