Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177056.4 | NP_796030.1 | 2 | 408 | ||
GenBank mRNA | AK038957.1 | - | 2 | 369 | ||
AK082334.1 | - | 1 | 668 | |||
AK158863.1 | - | 2 | 882 | |||
BC059868.1 | - | 2 | 373 | 6809137 | ||
CJ147397.1 | - | 2 | 408 |