Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177092.4 | NP_796066.1 | 7 | 686 | ||
GenBank mRNA | AK052456.1 | - | 4 | 299 | ||
AK086975.1 | - | 7 | 653 | |||
AK137163.1 | - | 7 | 653 | |||
BC096447.1 | - | 7 | 567 | 6310686 |