Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038624.1 | NP_001033713.1 | 5 | 582 | ||
NM_178780.3 | NP_848895.2 | 5 | 582 | |||
GenBank mRNA | AK053760.1 | - | 5 | 563 | ||
AK082275.1 | - | 5 | 563 | |||
AK134663.1 | - | 5 | 564 | |||
AK138641.1 | - | 5 | 578 | |||
AK149334.1 | - | 5 | 570 | |||
BC059258.1 | - | 4 | 324 | 6408852 |