Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610524H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181075.3 | NP_851420.1 | 2 | 276 | ||
GenBank mRNA | AK012147.1 | - | 2 | 265 | ||
AK148078.1 | - | 2 | 251 | |||
BC089033.1 | - | 2 | 256 | |||
BC096574.1 | - | 2 | 267 | 1495259 | ||
BY090347.1 | - | 2 | 276 |
Gene Symbol: 1500011B03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161363.1 | - | 2 | 287 | ||
BC061238.1 | - | 2 | 269 |