Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081308.2 | NP_001074777.1 | 17, 18 | 2424 | ||
NM_001199685.1 | NP_001186614.1 | 18, 19 | 2532 | |||
NM_183306.2 | NP_899129.2 | 18, 19 | 2509 | |||
GenBank mRNA | AK039665.1 | - | 5, 6 | 1082 | ||
AK217809.1 | - | 1, 2 | 43 | |||
BC158072.1 | - | 18, 19 | 2474 | 9088167 | ||
CK625948.1 | - | 3, 4 | 481 |