Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177876.4 | NP_808544.1 | 4 | 655 | ||
GenBank mRNA | AK149893.1 | - | 4 | 622 | ||
AK166359.1 | - | 4 | 636 | |||
AK172114.1 | - | 4 | 616 | |||
BC026744.1 | - | 4 | 656 | 4163734 |