Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177391.4 | NP_796365.1 | 2 | 826 | ||
GenBank mRNA | AK083325.1 | - | 2 | 801 | ||
AK090060.1 | - | 2 | 922 | |||
BC116256.1 | - | 1 | 594 | 40093730 | ||
BC116257.1 | - | 1 | 591 | 40093731 |