Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029548.4 | NP_083824.1 | 5 | 611 | ||
GenBank mRNA | AB112925.1 | - | 5 | 578 | ||
AB158403.1 | - | 5 | 619 | |||
BC127960.1 | - | 5 | 582 | 40134813 | ||
DV651301.1 | - | 4 | 458 | |||
DV660756.1 | - | 6 | 703 |