Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024624.2 | NP_001019795.1 | 12 | 1748 | ||
GenBank mRNA | AK052380.1 | - | 12 | 1745 | ||
AK139220.1 | - | 12 | 1720 | |||
BC042705.1 | - | 3 | 722 |