Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207525.3 | NP_997408.2 | 1 | 158 | ||
GenBank mRNA | AK052774.1 | - | 1 | 165 | ||
AK133480.1 | - | 144 | ||||
AK154917.1 | - | 164 | ||||
AK154960.1 | - | 159 | ||||
AK164755.1 | - | 1 | 159 | |||
AK165767.1 | - | 1 | 189 | |||
AK170977.1 | - | 1 | 207 | |||
BC094601.1 | - | 1 | 194 | 6404450 | ||
BC132309.1 | - | 1 | 143 | 40130586 | ||
BC132311.1 | - | 1 | 143 | 40130588 |