Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 4 | 497 | ||
GenBank mRNA | AK080983.1 | - | 638 | |||
AK088196.1 | - | 4 | 510 | |||
AK151326.1 | - | 483 | ||||
AK151705.1 | - | 4 | 507 | |||
AK170042.1 | - | 507 | ||||
BC002270.1 | - | 4 | 497 | 3496088 |
Gene Symbol: Gm6194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BY705843.1 | - | 1 | 159 | ||
CF106116.1 | - | 1 | 107 | |||
CJ047045.1 | - | 1 | 165 |