Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011684.2 | NP_001011684.3 | 2, 3 | 270 | ||
GenBank mRNA | AB164466.1 | - | 2, 3 | 270 | ||
BC138922.1 | - | 2, 3 | 213 | 8861944 | ||
BU609935.1 | - | 2, 3 | 216 |