Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133237.3 | NP_573500.2 | 5 | 1599 | ||
GenBank mRNA | AK156319.1 | - | 5 | 1600 | ||
AK157407.1 | - | 167 | ||||
AK172004.1 | - | 1603 | ||||
AK190166.1 | - | 2 | 216 | |||
AK192811.1 | - | 2 | 216 | |||
AK194547.1 | - | 2 | 216 | |||
AK195006.1 | - | 2 | 216 | |||
AK195220.1 | - | 2 | 216 | |||
AK195944.1 | - | 2 | 216 | |||
AK198121.1 | - | 2 | 216 | |||
AK198472.1 | - | 2 | 215 | |||
AK198593.1 | - | 2 | 216 | |||
AK198892.1 | - | 2 | 216 | |||
AK202397.1 | - | 2 | 216 | |||
AK202718.1 | - | 2 | 216 | |||
AK204993.1 | - | 2 | 216 | |||
BC141269.1 | - | 6 | 1373 | 9055881 | ||
BC141270.1 | - | 6 | 1419 | 9055882 |