Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022653.4 | NP_073144.3 | 10, 11 | 1830 | ||
GenBank mRNA | AF314187.1 | - | 10, 11 | 1788 | ||
AK028609.1 | - | 10, 11 | 1764 | |||
BC031175.1 | - | 2, 3 | 341 | |||
BC031722.1 | - | 10, 11 | 1796 | 4019169 | ||
BC039651.1 | - | 2, 3 | 341 |