Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015794.1 | NP_056609.1 | 6 | 2287 | ||
GenBank mRNA | AF176529.1 | - | 6 | 1007 | ||
AK154497.1 | - | 6 | 796 | |||
BC058120.1 | - | 3 | 345 | 6810624 | ||
BC138330.1 | - | 3 | 323 | 8861350 | ||
BC138331.1 | - | 3 | 323 | 8861351 |