Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016968.4 | NP_058664.2 | 1 | 558 | ||
GenBank mRNA | AB038696.1 | - | 1 | 539 | ||
AK144435.1 | - | 1 | 556 | |||
AK158791.1 | - | 1 | 252 | |||
BC046316.1 | - | 1 | 517 | 6314266 |