Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267620.1 | NP_001254549.1 | 2, 3 | 623 | ||
NM_026187.5 | NP_080463.3 | 2, 3 | 623 | |||
GenBank mRNA | AK004948.2 | - | 2, 3 | 591 | ||
AK147915.1 | - | 1, 2 | 2134 | |||
AK156255.1 | - | 2, 3 | 587 | |||
AK157405.1 | - | 2, 3 | 587 | |||
BC039910.1 | - | 2, 3 | 586 | |||
BC051449.1 | - | 2, 3 | 603 | |||
BC071240.1 | - | 2, 3 | 522 | |||
BC150879.1 | - | 2, 3 | 573 | 9087790 | ||
DV062487.1 | - | 2, 3 | 647 |