Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144811.3 | NP_659060.1 | 1, 2 | 197 | ||
GenBank mRNA | AB221539.1 | - | 1, 2 | 71 | ||
AK045714.1 | - | 1, 2 | 191 | |||
AY453793.1 | - | 1, 2 | 187 | |||
BC021398.1 | - | 1, 2 | 213 | 5066655 | ||
BC042657.1 | - | 1, 2 | 319 |