Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144811.3 | NP_659060.1 | 4 | 361 | ||
GenBank mRNA | AK045714.1 | - | 4 | 355 | ||
BC021398.1 | - | 4 | 377 | 5066655 | ||
BC042657.1 | - | 4 | 483 | |||
BQ571764.1 | - | 2 | 128 |