Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vti1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293685.1 | NP_001280614.1 | 4 | 682 | ||
NM_001293686.1 | NP_001280615.1 | 4 | 682 | |||
NM_016862.4 | NP_058558.1 | 4 | 682 | |||
GenBank mRNA | AF035209.1 | - | 4 | 389 | ||
AF035823.1 | - | 4 | 387 | |||
AK004751.1 | - | 4 | 378 | |||
AK028646.1 | - | 4 | 388 | |||
AK030538.1 | - | 4 | 407 | |||
AK135463.1 | - | 4 | 445 | |||
AK148281.1 | - | 4 | 413 | |||
BC089321.1 | - | 4 | 389 | 6839437 |