Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016879.2 | NP_058575.2 | 3, 4 | 778 | ||
GenBank mRNA | AF021836.1 | - | 3, 4 | 762 | ||
AK009099.1 | - | 3, 4 | 777 | |||
AK028825.1 | - | 3, 4 | 778 |