Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281929.1 | NP_001268858.1 | 9, 10 | 1540 | ||
NM_001290451.1 | NP_001277380.1 | 9, 10 | 1312 | |||
NM_016886.4 | NP_058582.3 | 9, 10 | 1540 | |||
GenBank mRNA | AB022342.1 | - | 9, 10 | 1330 | ||
AB079072.1 | - | 9, 10 | 1276 | |||
AB102775.1 | - | 9, 10 | 1276 | |||
AB102776.1 | - | 9, 10 | 1276 | |||
AF483494.1 | - | 9, 10 | 1276 | |||
AF483495.1 | - | 9, 10 | 1276 | |||
AK135043.1 | - | 9, 10 | 1294 | |||
AK138615.1 | - | 9, 10 | 1445 | |||
AK157815.1 | - | 10, 11 | 1717 | |||
AK161163.1 | - | 9, 10 | 1360 | |||
AK220530.1 | - | 9, 10 | 1327 | |||
BC076584.1 | - | 9, 10 | 1404 | |||
BC086678.1 | - | 9, 10 | 1485 | 30668476 | ||
BC129855.1 | - | 9, 10 | 1330 | 40109751 | ||
BC129856.1 | - | 9, 10 | 1330 | 40109752 |