Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281929.1 | NP_001268858.1 | 11 | 2051 | ||
NM_001290451.1 | NP_001277380.1 | 11 | 1823 | |||
NM_016886.4 | NP_058582.3 | 11 | 2051 | |||
GenBank mRNA | AB022342.1 | - | 11 | 1841 | ||
AB079072.1 | - | 11 | 1787 | |||
AB102775.1 | - | 11 | 1787 | |||
AB102776.1 | - | 11 | 1787 | |||
AF483494.1 | - | 11 | 1787 | |||
AF483495.1 | - | 11 | 1787 | |||
AK054154.1 | - | 1 | 1456 | |||
AK135043.1 | - | 11 | 1805 | |||
AK138615.1 | - | 11 | 1956 | |||
AK157815.1 | - | 12 | 2228 | |||
AK161163.1 | - | 11 | 1871 | |||
AK220530.1 | - | 11 | 1838 | |||
BC076584.1 | - | 11 | 1915 | |||
BC086678.1 | - | 11 | 1996 | 30668476 | ||
BC129855.1 | - | 11 | 1841 | 40109751 | ||
BC129856.1 | - | 11 | 1841 | 40109752 |