Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017472.4 | NP_059500.2 | 2 | 477 | ||
GenBank mRNA | AF062482.1 | - | 2 | 275 | ||
AK007798.1 | - | 2 | 288 | |||
AK013183.1 | - | 2 | 502 | |||
AK018790.1 | - | 2 | 110 | |||
AK145350.1 | - | 2 | 280 | |||
AK154728.1 | - | 2 | 280 | |||
AK189751.1 | - | 2 | 68 | |||
BC116885.1 | - | 2 | 321 | 40126204 | ||
BC116889.1 | - | 2 | 321 | 40126208 |