Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019421.3 | NP_062294.3 | 3 | 405 | ||
GenBank mRNA | AK021187.1 | - | 3 | 380 | ||
AK078151.1 | - | 3 | 406 | |||
AK088189.1 | - | 3 | 406 | |||
AK131655.1 | - | 3 | 411 | |||
AK140204.1 | - | 3 | 392 | |||
BC026888.1 | - | 3 | 359 | 3991556 | ||
KJ650205.1 | - | 3 | 325 | |||
KJ663715.1 | - | 3 | 283 |