Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018773.2 | NP_061243.1 | 11 | 1113 | ||
GenBank mRNA | AB014485.1 | - | 11 | 1030 | ||
AF051324.1 | - | 11 | 1028 | |||
AK076000.1 | - | 11 | 1058 | |||
AK144289.1 | - | 11 | 1050 | |||
AK204408.1 | - | 3 | 249 | |||
BC003711.1 | - | 11 | 1113 | 3599914 |