Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018762.1 | NP_061232.1 | 2 | 555 | ||
GenBank mRNA | AB007464.1 | - | 2 | 555 | ||
BC111106.1 | - | 2 | 550 | 40055578 |