Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207202.2 | NP_997085.2 | 10 | 1511 | ||
GenBank mRNA | BC053918.1 | - | 10 | 1476 | ||
BC139064.1 | - | 9 | 1287 | 8862086 | ||
BU505792.1 | - | 5 | 721 |