Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026842.4 | NP_081118.4 | 11 | 1925 | ||
NM_152234.2 | NP_689420.1 | 10 | 1841 | |||
GenBank mRNA | AK004183.1 | - | 4 | 1209 | ||
AK048534.1 | - | 10 | 1762 | |||
AK049298.1 | - | 11 | 1680 | |||
AK131766.1 | - | 4 | 537 | |||
AK165456.1 | - | 10 | 1806 | |||
AK172695.1 | - | 10 | 1810 | |||
AK179463.1 | - | 2 | 246 | |||
AK202633.1 | - | 2 | 189 | |||
BC010213.1 | - | 8 | 1268 | |||
BC026847.1 | - | 11 | 1908 | 4015641 | ||
BC027375.1 | - | 5 | 628 | |||
BC028857.1 | - | 10 | 1780 | 4507759 | ||
BC051098.1 | - | 10 | 1553 | |||
BC080667.1 | - | 11 | 1886 | 4935292 |