Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009935.2 | NP_001009935.1 | 3 | 691 | ||
NM_023719.2 | NP_076208.2 | 3 | 688 | |||
GenBank mRNA | AF173681.1 | - | 3 | 455 | ||
AF282826.1 | - | 3 | 409 | |||
AK082476.1 | - | 3 | 686 | |||
AK088870.1 | - | 3 | 686 | |||
AK089304.1 | - | 3 | 691 | |||
AK089351.1 | - | 3 | 689 | |||
AK089403.1 | - | 3 | 691 | |||
AK155517.1 | - | 3 | 689 | |||
AK171205.1 | - | 3 | 689 | |||
AK171312.1 | - | 3 | 689 | |||
AK171425.1 | - | 3 | 689 | |||
AK171456.1 | - | 3 | 686 | |||
AK171464.1 | - | 3 | 689 | |||
AK171527.1 | - | 3 | 689 | |||
AK192038.1 | - | 3 | 286 | |||
AK214578.1 | - | 3 | 263 | |||
BC011212.1 | - | 3 | 667 | 4193517 | ||
BC031850.1 | - | 3 | 666 | 3591421 |