Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019964.1 | NP_064348.1 | 1 | 558 | ||
GenBank mRNA | AB028856.1 | - | 1 | 558 | ||
AK006026.1 | - | 1 | 542 | |||
BC049591.1 | - | 1 | 536 | 6773959 | ||
BC061112.1 | - | 1 | 541 | 6772873 |
Gene Symbol: 1700031F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI647336.1 | - | 2 | 365 | ||
AK006579.1 | - | 2 | 326 | |||
BC099573.1 | - | 2 | 384 | |||
CN837050.1 | - | 387 |