Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019937.3 | NP_064321.2 | 9 | 1173 | ||
GenBank mRNA | AF159159.1 | - | 9 | 1126 | ||
AF467251.1 | - | 9 | 1167 | |||
BC007177.1 | - | 8 | 3161 | |||
BC037204.1 | - | 8 | 2800 | |||
BC069909.1 | - | 10 | 1257 | |||
BC094383.1 | - | 9 | 1109 | 30604933 | ||
BC117816.1 | - | 10 | 1066 |