Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020587.2 | NP_065612.2 | 1 | 190 | ||
GenBank mRNA | AK075769.1 | - | 1 | 158 | ||
AK077648.1 | - | 1 | 176 | |||
AK156989.1 | - | 1 | 125 | |||
BC019437.1 | - | 1 | 150 | 4018178 | ||
BY107258.1 | - | 1 | 191 |