Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam196b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025382.2 | NP_001020553.1 | 2 | 2214 | ||
GenBank mRNA | BC072605.1 | - | 3 | 1875 | 6831248 |